Skip to Content
Merck
All Photos(1)

Key Documents

341552

Sigma-Aldrich

Anti-Fibrinogen Rabbit pAb

liquid, ≥90% (IgG)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

Assay

≥90% (IgG)

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FIBCD1(84929)

General description

Anti-Fibrinogen, rabbit polyclonal, recognizes human fibrinogen. It is validated for use in ELISA, immunoelectrophoresis, and radial immunodiffusion.
Rabbit polyclonal antibody purified by ammonium sulfate precipitation and DEAE-cellulose ion exchange chromatography. Recognizes the fibrinogen protein.
Recognizes fibrinogen.

Immunogen

Human
human fibrinogen

Application

ELISA (1:4000)

Immunoelectrophoresis (see comments)

Radial Immunodiffusion (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Monospecific for fibrinogen as determined by a single arc by immunoelectrophoresis against whole human plasma. This antibody has also been reported to work for radial immunodiffusion. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irina D Pokrovskaya et al.
Research and practice in thrombosis and haemostasis, 4(1), 72-85 (2020-01-29)
State-of-the-art 3-dimensional (3D) electron microscopy approaches provide a new standard for the visualization of human platelet ultrastructure. Application of these approaches to platelets rapidly fixed prior to purification to minimize activation should provide new insights into resting platelet ultrastructure. Our
Ruth C Massey et al.
Infection and immunity, 70(10), 5339-5345 (2002-09-14)
Defining the role of Staphylococcus aureus adhesins in disease pathogenesis may depend on the use of bacteria grown in culture media that more closely reflect the human milieu than conventional broth. This study examined the functional effect on S. aureus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service