Skip to Content
Merck
All Photos(1)

Key Documents

324683

Sigma-Aldrich

Ellagic Acid, Dihydrate

A cell-permeable, reversible, potent antioxidant that has anti-mutagenic and anti-carcinogenic properties.

Synonym(s):

Ellagic Acid, Dihydrate, 4,4ʹ,5,5ʹ,6,6ʹ-Hexahydroxydiphenic Acid 2,6,2ʹ,6ʹ-Dilactone, TBBD, PRMT Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H6O8 · 2H2O
CAS Number:
Molecular Weight:
338.22
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 3547

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

slightly beige

solubility

1 M NaOH: 10 mg/mL
ethanol: 10 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

[o]1c2c3c4c([o][c](c3cc(c2O)O)=O)c(c(cc4[c]1=O)O)O

InChI

1S/C14H6O8/c15-5-1-3-7-8-4(14(20)22-11(7)9(5)17)2-6(16)10(18)12(8)21-13(3)19/h1-2,15-18H

InChI key

AFSDNFLWKVMVRB-UHFFFAOYSA-N

General description

A cell-permeable, reversible, and potent antioxidant that has anti-mutagenic and anti-carcinogenic properties. Acts as a selective and ATP-competitive inhibitor of CK2 (IC50 = 40 nM). Moderately inhibits DNA topoisomerases I and II, Lyn, PKA catalytic subunit, Lyk, GSK-3, PKC, and FGR (IC50 = 1.8, 2.1, 2.9, 3.5, 4.3, 7.5, 8.0, and 9.4 µM, respectively). Only minimally inhibits DYRK1a, CSK, RET, Flt3, and NPM-ALK (IC50 >40 µM). Shown to act as an uncompetitive inhibitor of arginine methyltransferase CARM1 and block histone H3R17 methylation.
A cell-permeable, reversible, potent antioxidant that has anti-mutagenic and anti-carcinogenic properties. Acts as a selective and ATP-competitive inhibitor of CK2 (IC50 = 40 nM). Moderately inhibits DNA topoisomerases I and II, Lyn, PKA catalytic subunit, Lyk, GSK-3, PKC, and FGR (IC50 = 1.8, 2.1, 2.9, 3.5, 4.3, 7.5, 8.0, and 9.4 µM, respectively). Only minimally inhibits DYRK1a, CSK, RET, Flt3, and NPM-ALK (IC50 >40 µM). Shown to act as an uncompetitive inhibitor of arginine methyltransferase CARM1 and block histone H3R17 methylation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CDK2
Product competes with ATP.
Reversible: yes
Target IC50: 40 nM against CK2; 1.8, 2.1, 2.9, 3.5, 4.3, 7.5, 8.0, and 9.4 µM, against DNA topoisomerases I and II, Lyn, PKA catalytic subunit, Lyk, GSK-3, PKC, and FGR, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Preparation Note

For complete solubilization in ethanol, slight heating may be required.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Ethanol stock solutions are stable for up to 1 week at -20°C. Alkaline solutions are unstable and should be prepared just prior to use.

Other Notes

Selvi, B.R., et al. 2009, J. Biol. Chem. In press.
Cozza, G., et al. 2006. J. Med. Chem.49, 2363.
Wang, B.H., et al. 1998. Planta Med. 64, 195.
Constantinou, A., et al. 1995. Nutr. Cancer23, 121.
Hickey, M.J., et al. 1995. Biochem. Soc. Trans.23, 607s.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Are there any studies with ellagic acid and how it interferes with the spread of cancer?

    1 answer
    1. Ellagic acid shows significant promise as an anticancer agent through its ability to inhibit cancer cell proliferation, induce apoptosis, and prevent metastasis by modulating various molecular pathways.
      For more detailed insights refer to the following studies:
      https://pubmed.ncbi.nlm.nih.gov/38002335/
      https://pmc.ncbi.nlm.nih.gov/articles/PMC4069806/
      https://www.sciencedirect.com/science/article/pii/S1878535224001278

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service