Skip to Content
Merck
All Photos(1)

Key Documents

311155

Sigma-Aldrich

Dansyl Chloride

Reagent used for fluorescent labeling of proteins, N-terminal amino acids, and amines.

Synonym(s):

Dansyl Chloride, 5-(Dimethylamino)-napthalene-1-sulfonyl Chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12ClNO2S
CAS Number:
Molecular Weight:
269.75
MDL number:
UNSPSC Code:
12161703
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

yellow-orange

solubility

DMF: soluble
chloroform: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(Cl)c1c2c(c(ccc2)N(C)C)ccc1

InChI

1S/C12H12ClNO2S/c1-14(2)11-7-3-6-10-9(11)5-4-8-12(10)17(13,15)16/h3-8H,1-2H3

InChI key

XPDXVDYUQZHFPV-UHFFFAOYSA-N

General description

Reagent used for fluorescent labeling of proteins, N-terminal amino acids, and amines.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Reagent used for fluorescent labeling of proteins, N-terminal amino acids, and amines
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Corrosive (L)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Vera, J.C., et al. 1988. Anal. Biochem. 174, 38.
Gehring, U., et al. 1968. Eur. J. Biochem. 6, 281.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service