Skip to Content
Merck
All Photos(2)

Documents

239824

Sigma-Aldrich

CXCR7 Agonist, VUF11207

The CXCR7 Agonist, VUF11207 controls the biological activity of CXCR7. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

CXCR7 Agonist, VUF11207, (E)-N-(3-(2-fluorophenyl)-2-methylallyl)-3,4,5-trimethoxy-N-(2-(1-methylpyrrolidin-2-yl)ethyl)benzamide, RDC1 agonist, CMKOR1 Agonist, CXCR-7 Agonist, CXC-R7 Agonist, RDC-1 Agonist, GPR159 Agonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H35FN2O4
CAS Number:
Molecular Weight:
470.58
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(N(CCC1N(C)CCC1)C/C(C)=C/C2=C(F)C=CC=C2)C3=CC(OC)=C(OC)C(OC)=C3

General description

A styrene-amide derivative that acts as a high affinity and high potency ligand of CXCR7 chemokine receptor (pKi = 8.1) and induces the recruitment of β-arrestin2 and subsequent interenalization of CXCR7. Exposure of HEK293 T cells transiently transfected with cDNA encoding both CXCR7-RLuc and β-arrestin 2-YFP to VUF11207 is shown to induce a dose-dependent increase in bioluminescence resonance energy transfer (BRET) signal (EC50 = 1.6 nM) indicating its agonist role in β-arrestin2 is recruited to the active receptor.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CXCR7
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Wijtmans, M., et al. 2011, Eur J Med Chem51, 184.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service