Skip to Content
Merck
All Photos(1)

Documents

227036

Sigma-Aldrich

Cholera Toxin

from Vibrio cholerae, ≥95% (SDS-PAGE), solid, A-B type toxin, Calbiochem

Synonym(s):

Cholera Toxin, Vibrio cholerae, Type Inaba 569B, Azide Free

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.51

product name

Cholera Toxin, Vibrio cholerae, Type Inaba 569B, Azide Free, Cholera toxin Inaba 569B, CAS 9012-63-9, has a single A subunit surrounded by five polypeptides chains of the B subunits. The A subunit catalyzes the ADP-ribosylation of the α-subunit of G-proteins.

Quality Level

Assay

≥95% (SDS-PAGE)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

shipped in

ambient

storage temp.

2-8°C

General description

Useful for studies involving adenylate cyclase, AMP, and related membrane-transport phenomena. Consists of a single A subunit surrounded by five polypeptide chains of the B subunits. The A subunit catalyzes the ADP-ribosylation of the α-subunit of G-proteins, reducing intrinsic GTPase activity and activating the α-subunit. The B subunits are responsible for the attachment of the native toxin on the cell surface.
Useful for studies involving adenylate cyclase, AMP, and related membrane-transport phenomena. Consists of a single A subunit surrounded by five polypeptide chains of the B subunits. The A subunit catalyzes the ADP-ribosylation of the α-subunit of G-proteins, reducing intrinsic GTPase activity and activating the α-subunit. The B subunits are responsible for the attachment of the native toxin on the cell surface..
Useful for tissue culture applications.

Biochem/physiol Actions

Cell permeable: no
Primary Target
α-subunit of G-proteins
Product does not compete with ATP.
Reversible: no

Packaging

Please refer to vial label for lot-specific concentration.
yes

Warning

Toxicity: Highly Toxic (H)

Physical form

Lyophilized from 200 mM NaCl, 50 mM Tris, 1 mM EDTA, pH 7.5.

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Aqueous stock solutions are stable for up to 6 months at 4°C under sterile conditions.
Please refer to the vial label for lot-specific reconstitution. Note: cholera toxin tends to breakdown into its A and B subunits when the solution is frozen or with vigorous shaking during reconstitution, resulting in damage to the toxin protein and concomitant loss of biological activity. Swirl bottles gently during reconstitution. Avoid vigorous pipetting of solutions that would lead to foaming. Heating of cholera toxin in excess of 40-50°C results in aggregation and subsequent breakdown into its A and B subunits.

Analysis Note

Binding activity is assessed by hemagglutination assay using trisialoganglioside fixed sheep red blood cells.
single band by disc gel electrophoresis

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Gilman, A.G. 1987. Annu. Rev.Biochem.56, 615.

Legal Information

Not available for sale outside of the United States.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service