Skip to Content
Merck
All Photos(1)

Key Documents

217720

Sigma-Aldrich

Cdk1/5 Inhibitor

The Cdk1/5 Inhibitor, also referenced under CAS 40254-90-8, controls the biological activity of Cdk1/5. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Cdk1/5 Inhibitor, 3-Amino-1H-pyrazolo[3,4-b]quinoxaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N5
CAS Number:
Molecular Weight:
185.19
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C9H7N5/c10-8-7-9(14-13-8)12-6-4-2-1-3-5(6)11-7/h1-4H,(H3,10,12,13,14)

InChI key

DWHVZCLBMTZRQM-UHFFFAOYSA-N

General description

A 3-amino substituted pyrazoloquinoxaline compound that acts as a selective inhibitor of cyclin-dependent kinases 1 and 5 (Cdks; IC50 = 0.6 µM and 0.4 µM for Cdk1/cyclin B and Cdk5/p25, respectively). Also shown to inhibit GSK-3β with (IC50 = 1 µM). Does not inhibit Cdc25 phosphatase activity (IC50 >10 µM).
A selective inhibitor of cyclin-dependent kinases 1 and 5 (Cdk1/Cdk5) (IC50 = 600 nM for Cdk1/cyclin B and 400 nM for Cdk5/p25). Also shown to inhibit glycogen synthase kinase-3β (GSK-3β) (IC50 = 1 µM). Does not inhibit Cdc25 phosphatase activity (IC50 >10 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Cdk1/cyclin B, Cdk5/p25
Product does not compete with ATP.
Reversible: no
Target IC50: 0.6 µM and 0.4 µM for Cdk1/cyclin B and Cdk5/p25, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ortega, M.A., et al. 2002. Bioorg. Med. Chem.10, 2177.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service