Skip to Content
Merck
All Photos(1)

Documents

20-284

Millipore

Catch and Release Denaturing Elution Buffer, 1X

Denaturing Elution Buffer designed for use with the Catch & Release reversible immunoprecipitation system to recover captured immunocomplex from Catch & Release Spin Columns.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.22

Quality Level

form

liquid

packaging

pkg of 4 mL

manufacturer/tradename

Catch And Release
Upstate®

technique(s)

immunoprecipitation (IP): suitable

shipped in

wet ice

Application

Designed for use with the Catch and Release reversible immunoprecipitation system to recover captured immunocomplex from Catch and Release Spin Columns

Quality

routinely evaluated as elution buffer for Catch and Release reversible immunoprecipitatoin kit (Cat#17-500)

Storage and Stability

Stable for 1 year at 4°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service