Skip to Content
Merck
All Photos(1)

Documents

200100

Sigma-Aldrich

Benzyl-2-acetamido-2-deoxy-α-D-galactopyranoside

≥98% (TLC), solid, O-linked glycosylation inhibitor, Calbiochem®

Synonym(s):

Benzyl-2-acetamido-2-deoxy-α-D-galactopyranoside, Benzyl-α-GalNAc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO6
CAS Number:
Molecular Weight:
311.33
MDL number:
UNSPSC Code:
12352200

product name

Benzyl-2-acetamido-2-deoxy-α-D-galactopyranoside, Used as an inhibitor of O-linked glycosylation in a variety of cell lines.

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

hot ethanol: 1 mg/mL
DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

CC(N[C@H]([C@H]([C@H]1O)O)[C@H](O[C@@H]1CO)OCC2=CC=CC=C2)=O

InChI

1S/C15H21NO6/c1-9(18)16-12-14(20)13(19)11(7-17)22-15(12)21-8-10-5-3-2-4-6-10/h2-6,11-15,17,19-20H,7-8H2,1H3,(H,16,18)/t11-,12-,13+,14-,15+/m1/s1

InChI key

SKOZFDIGKDPQBO-QMIVOQANSA-N

General description

Used as an inhibitor of O-linked glycosylation in a variety of cell lines. Also inhibits 2,3(O)-sialyltransferase and disrupts glycoprotein targeting in HT-29 cells. Substrate for N-acetyl-β-D-glucosaminyltransferase.
Used as an inhibitor of O-linked glycosylation in a variety of cell lines. It has also been used to inhibit 2,3(O)-sialyltransferase and disrupt glycoprotein targeting in HT-29 cells. Substrate for N-acetyl-β-D-glucosaminyltransferase.

Biochem/physiol Actions

Cell permeable: no
Primary Target
o-linked glycosylation
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Other Notes

Huet, G., et al. 1998. J. Cell Biol. 141, 1311.
Kuan, S.-F., et al. 1989. J. Biol. Chem. 264, 19271.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service