Skip to Content
Merck
All Photos(2)

Key Documents

197227

Sigma-Aldrich

BCRP Inhibitor III, YHO-13177

The BCRP Inhibitor III, YHO-13177 controls the biological activity of BCRP.

Synonym(s):

BCRP Inhibitor III, YHO-13177, ( Z)-2-(3,4-Dimethoxyphenyl)-3-(5-(4-hydroxypiperidin-1-yl)thiophen-2-yl)acrylonitrile, (Z)-2-(3,4-Dimethoxyphenyl)-3-(5-(4-hydroxypiperidin-1-yl)thiophen-2-yl)acrylonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N2O3S
CAS Number:
Molecular Weight:
370.47
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

OC(CC1)CCN1C(S2)=CC=C2/C=C(C#N)/C3=CC(OC)=C(OC)C=C3

General description

A cell-permeable acrylonitrile compound that potently inhibits breast cancer resistance protein (BCRP/ABCG2)-mediated drug export and reverses BCRP-mediated drug resistance in vitro (half maximal in the range 0.01 - 1 µM) without altering the BCRP expression levels (in ≤ 6 hrs.). Shown to selectively enable the intracellular accumulation of anticancer drugs and potentiate the cytotoxicity of BCRP-expressing carcinoma cells with no effect either on the P-glycoprotein/ABCB1- and MDRP1-mediated export or on the drug sensitivity of parental cells (IC50 >10 µM in A549 cells). An in vivo bioavailable and readily water soluble prodrug of YHO-13177, BCRP Inhibitor IV, YHO-13351 is also available (Cat. No. 197228).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Yamazaki, R., et al. 2011. Mol. Cancer Ther.10, 1252.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service