Skip to Content
Merck
All Photos(1)

Key Documents

1.93417

Supelco

L(+)-Tartaric acid

EMPLURA®

Synonym(s):

L(+)-Tartaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH(OH)CH(OH)COOH
CAS Number:
Molecular Weight:
150.09
MDL number:
UNSPSC Code:
12352106
Pricing and availability is not currently available.

product line

EMPLURA®

storage temp.

room temp

SMILES string

[O-]C(=O)C(O)C(O)C(=O)[O-].[H+].[H+]

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)

InChI key

FEWJPZIEWOKRBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93417.0521: 500 g, Minimum order quantity: 20
1.93417.9051: 50 kg, Minimum order quantity: 1

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service