Skip to Content
Merck
All Photos(1)

Key Documents

1.93310

Supelco

Ammonium cerium(IV) nitrate

for analysis EMPARTA® ACS

Synonym(s):

Ammonium cerium(IV) nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2[Ce(NO3)6]
CAS Number:
Molecular Weight:
548.22
MDL number:
UNSPSC Code:
12352302
Pricing and availability is not currently available.

grade

ACS reagent

storage temp.

room temp

SMILES string

[Ce+4].[N+](=O)([O-])O.[N+](=O)([O-])O.[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].N.N

InChI

1S/Ce.2HNO3.4NO3.2H3N/c;6*2-1(3)4;;/h;2*(H,2,3,4);;;;;2*1H3/q+4;;;4*-1;;

InChI key

WIBGOERAEYJBOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93310.0121: 100 g, Minimum order quantity: 20

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service