Skip to Content
Merck
All Photos(1)

Key Documents

1.93208

Supelco

Potassium thiocyanate

for analysis EMPARTA® ACS

Synonym(s):

Potassium thiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CKNS
CAS Number:
Molecular Weight:
97.18
MDL number:
UNSPSC Code:
12352302

grade

ACS reagent

storage temp.

room temp

InChI

1S/CHNS.K/c2-1-3;/h3H;/q;+1/p-1

InChI key

ZNNZYHKDIALBAK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93208.0521: 500 g, Minimum order quantity: 1

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service