Skip to Content
Merck
All Photos(1)

Key Documents

176880

Sigma-Aldrich

Anisomycin, Streptomyces griseolus

Strongly activates stress-activated protein kinases (JNK/SAPK) and p38 MAP kinase in mammalian cells.

Synonym(s):

Anisomycin, Streptomyces griseolus, 2-( p-Methoxybenzyl)-3,4-pyrrolidinediol-3-acetate, 2-(p-Methoxybenzyl)-3,4-pyrrolidinediol-3-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO4
CAS Number:
Molecular Weight:
265.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 100 mg/mL
methanol: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C14H19NO4/c1-9(16)19-14-12(15-8-13(14)17)7-10-3-5-11(18-2)6-4-10/h3-6,12-15,17H,7-8H2,1-2H3/t12-,13+,14+/m0/s1

InChI key

YKJYKKNCCRKFSL-BFHYXJOUSA-N

General description

Activates p54 and MAP kinases. Involved in the activation of stress-activated protein kinases (SAPKs). Synergizes with growth factors to induce c-fos and c-jun by acting as a potent signaling agonist. An inhibitor of protein synthesis at the translation step. Also known to induce apoptosis in the human monoblastoid cell line, U937.
Strongly activates stress-activated protein kinases (JNK/SAPK) and p38 MAP kinase in mammalian cells. Synergizes with growth factors to induce c-fos and c-jun by acting as a potent signaling agonist. An inhibitor of protein synthesis at the translation step. Also known to induce apoptosis in the human monoblastoid cell line, U937.

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution store in the refrigerator (4°C). DMSO stock solutions are stable for up to 6 months at 4°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Hazzalin, C.A., et al. 1998. Mol. Cell. Biol. 18, 1844.
Kardalinou, E., et al. 1994. Mol. Cell. Biol.14, 1066.
Kyriakis, J.M., et al. 1994. Nature 369, 156.
Sanchez, I., et al. 1994. Nature 372, 794.
Shinohara, K., and Oka, T. 1994. NeuroReport 5, 2201.
Kochi, S. K., and Collier, R.J. 1993. Exp. Cell Res.208, 296.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James L McLellan et al.
Bio-protocol, 14(5), e4952-e4952 (2024-03-11)
The Plasmodium parasites that cause malaria undergo an obligate, asymptomatic developmental stage in the host liver before initiating the symptomatic blood-stage infection. The parasite liver stage is a key intervention point for antimalarial chemoprophylaxis: successful targeting of liver-stage parasites prevents
Madeleine Scharf et al.
Molecular and cellular biology, 33(13), 2586-2602 (2013-04-24)
The mitogen-activated protein kinase (MAPK)-activated protein kinases 2 and 3 (MK2/3) represent protein kinases downstream of the p38 MAPK. Using MK2/3 double-knockout (MK2/3(-/-)) mice, we analyzed the role of MK2/3 in cross-striated muscle by transcriptome and proteome analyses and by
James L McLellan et al.
Life science alliance, 7(6) (2024-04-05)
Increasing numbers of antimalarial compounds are being identified that converge mechanistically at inhibition of cytoplasmic translation, regardless of the molecular target or mechanism. A deeper understanding of how their effectiveness as liver stage translation inhibitors relates to their chemoprotective potential
Yuan Zhang et al.
Redox biology, 69, 103005-103005 (2023-12-28)
Major depressive disorder (MDD) is a devastating condition. Although progress has been made in the past seven decades, patients with MDD continue to receive an inadequate treatment, primarily due to the late onset of first-line antidepressant drugs and to their
Chunchu Deng et al.
Journal of cell science, 134(22) (2021-10-21)
In neurons, the endoplasmic reticulum (ER) forms a highly dynamic network that enters axons and presynaptic terminals and plays a central role in Ca2+ homeostasis and synapse maintenance; however, the underlying mechanisms involved in regulation of its dynamic remodeling as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service