Skip to Content
Merck
All Photos(3)

Key Documents

17-10260

Sigma-Aldrich

ChIPAb+ N-CoR Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Nuclear receptor corepressor 1, N-CoR, N-CoR1, Retinoid X receptor-interacting protein 13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+JMJD1C set includes the JMJD1C antibody, a Normal Rabbit IgG, and control primers which amplify a 110 bp region of ChIP Primers, human β-globin. The JMJD1C and negative controls are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of JMJD1C -associated chromatin.
In the absence of ligands, the corepressor N-CoR represses nuclear hormone receptor-mediated gene transcription. In the presence of ligands, these hormone receptors activate gene expression and play critical roles in development, growth, and homeostasis. N-Cor-mediated transcriptional repression is facilitated through the formation of histone deacetylase (HDACs) N-CoR complexes or by conjugation from small ubiquitin-like modifier 1 (SUMO-1). It is shown that the SANT1 domain of N-CoR serves as a binding region for SUMO-E2 conjugating enzyme Ubc9 and SUMO-E3 ligase Pias1, which is necessary for N-CoR-mediated repression of respective reporter genes. SUMOylation-deficient N-COR knockout mice display a signification reduction in the repression of gene transcription and result in defects involving the differentiation and development of neural cells into erythrocytes and thymocytes.

Immunogen

KLH-conjugated linear peptide corresponding to a region between amino acids 1510 and 1540 of the N-CoR protein.

Application

Chromatin Immunoprecipitation:
Representative lot data.
Sonicated chromatin prepared from untreated or UV treated (6 hrs, 50 joules/m2.) U2OS cells (3 X 10E6 cell equivalents per IP) was subjected to chromatin immunoprecipitation using using 3 µg of either Normal Rabbit IgGor 3 µg of Anti-N-CoR and the Magna ChIP A/G Kit (Cat. # 17-10085). Successful immunoprecipitation of N-CoR associated DNA fragments was verified by qPCR using ChIP Primers, p21. (Figure 2). Data is presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.
Western Blot Analysis:
Representative lot data.
RAW 264.7 cell lysate was probed with Anti-N-CoR (1 µg/mL). Proteins were visualized using a Donkey Anti-Rabbit IgG secondary antibody conjugated to HRP and a chemiluminescence detection system.
Arrow indicates Anti-N-CoR (~270 kDa). (Figure 3).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Hormones & Receptors
This ChIPAb+ N-CoR -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Packaging

25 assays per set. Recommended use: 3 µg of antibody per chromatin immuno-precipitation (dependent upon biological context).

Quality

Chromatin Immunoprecipitation:
Representative lot data.
Sonicated chromatin prepared from U2OS cells (3 X 10E6 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 3 µg of either Normal Rabbit IgGor 3 µg of Anti-N-CoR and the Magna ChIP® A/G Kit (Cat. # 17-10085). Successful immunoprecipitation of N-CoR associated DNA fragments was verified by qPCR using ChIP Primers, p21flanking an Sp1 binding site in the human p21 promoter. (Figure 1).
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Target description

~270 kDa observed. Uniprot describes 2 isoforms produced by alternative splicing at MW of 266 kDa and 270 kDa

Physical form

Affinity purified
Components:
Anti-N-CoR (Rabbit Polyclonal). One vial containing 75 µg of affinity purified polyclonal in buffer containing 0.1M Tris-glycine (pH 7.4) and 150 mM NaCl with 0.05% sodium azide before the addition of 30% glycerol. Store at -20°C.
Concentration: 0.7 mg/mL
Normal Rabbit IgG One vial containing 125 µg of Rabbit IgG in 125 µL of storage buffer containing 0.05% sodium azide. Store at -20°C.
ChIP Primers, p21. One vial containing 75 μL of each primer (5 μM) specific for the human p21 (WAF1/CIP1/CDKN1A) promoter. Store at
-20°C.
FOR: CCC ACA GCA GAG GAG AAA GAA
REV: CTG GAA ATC TCT GCC CAG ACA

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Includes normal rabbit IgG and primers specific for human p21.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Underhill et al.
The Journal of biological chemistry, 275(51), 40463-40470 (2000-10-03)
Transcriptional silencing by many transcription factors is mediated by the nuclear receptor corepressor (N-CoR). The mechanism by which N-CoR represses basal transcription involves the direct or indirect recruitment of histone deacetylases (HDACs). We have isolated two multiprotein N-CoR complexes, designated
Yan-Yun Liu et al.
Scientific reports, 11(1), 24105-24105 (2021-12-18)
Thyroid hormone (TH) and thyroid hormone receptor (THR) regulate stem cell proliferation and differentiation during development, as well as during tissue renewal and repair in the adult. THR undergoes posttranslational modification by small ubiquitin-like modifier (SUMO). We generated the THRA
G E Muscat et al.
Nucleic acids research, 26(12), 2899-2907 (1998-06-05)
Repression of transcription by the classical nuclear receptors (e.g. TR, RAR), the orphan nuclear receptors (e.g. Rev-erbAalpha/beta), Mxi-1 and Mad bHLH-zip proteins and the oncoproteins PLZF and LAZ3/BCL6 is mediated by the corepressors N-CoR and SMRT. The interaction of the
Laura Mikkonen et al.
Endocrinology, 154(2), 698-708 (2012-12-29)
Properly functioning adipose tissue is essential for normal insulin sensitivity of the body. When mice are kept on high-fat diet (HFD), adipose tissue expands, adipocytes increase in size and number, and the mice become obese. Many of these changes are
Jens Tiefenbach et al.
Molecular biology of the cell, 17(4), 1643-1651 (2006-01-20)
In the absence of ligands the corepressor N-CoR mediates transcriptional repression by some nuclear hormone receptors. Several protein-protein interactions of N-CoR are known, of which mainly complex formation with histone deacetylases (HDACs) leads to the repression of target genes. On

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service