Skip to Content
Merck
All Photos(1)

Documents

1.66509

Millipore

Sabouraud Dextrose Agar

with Lecithin and Tween® – 90mm ICR Settle plate, irradiated, triple packed, for environmental monitoring (Isolator and Clean room)

Synonym(s):

SDA, ICR Settle plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106227

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate

feature

2D bar code
ready-to-use

packaging

pkg of (20 Plates per box)

D

90 mm

pH

(7.3 (in H2O))

application(s)

air monitoring
microbiology
pharmaceutical

storage temp.

15-25°C

suitability

(Yeasts and Molds)

General description

Sabouraud Dextrose Agar + LT - ICR settle plate is a selective media for the determination of the total count of yeasts and molds. Settle plates are used for active or passive air monitoring as well for fingerprints of personnel. Sabouraud Dextrose Medium contains digests of animal tissues (peptones) which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is added as the energy and carbon source and low pH favours fungal growth. In addition, Lecithin and polysorbate 80 (Tween®) are neutralizers to inactivate residues of disinfectants.

Application

Sabouraud Dextrose Agar (SDA) is a selective medium for cultivation and isolation of yeasts and molds. The media is used for environmental monitoring by determining the yeast and mold count in air by active or passive air monitoring as well as fingerprints of personnel.

Packaging

1665090020-CN: 20 Plates per box

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service