Skip to Content
Merck
All Photos(1)

Key Documents

1590-OP

Millipore

Acrylamide/Bis-acrylamide

OmniPur® Grade, 40%, premixed powder, 19:1 ratio, Suitable for nucleic acids and protein separations and sequencing.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
Pricing and availability is not currently available.

Quality Level

form

free flowing powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

storage temp.

10-30°C

General description

Suitable for nucleic acids and protein separations and sequencing. Molecular biology grade. Premixed powder offers the convenience of three standard ratios of acrylamide to bis-acrylamide for convenience and safety. Reconstitute with deionized water to a final volume of 100 ml for the 40 g size and 500 ml for the 200 g size; yields a 40% working solution of 29: 1 ratio.

Specifications:
DNase: None detected
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service