Skip to Content
Merck
All Photos(1)

Key Documents

14-779-M

Sigma-Aldrich

MKP5, active

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

Quality Level

mol wt

Mw 20.6 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (phosphatase)

NCBI accession no.

General description

Dual specificity protein phosphatases inactivate their target kinases by dephosphorylating both the
phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the MAPK superfamily
(MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members ofthis family of dual specificity phosphatases show distinct substrate specificities for MAPKs, different tissue
distribution and subcellular localization, and different modes of inducibility of their expression by extracellular
stimuli. This gene product binds to and inactivates p38 and SAPK/JNK, but not MAPK/ERK. Its subcellular localization
is unique; it is evenly distributed in both the cytoplasm and the nucleus. This gene is widely expressed in various
tissues and organs, and its expression is elevated by stress stimuli. Three transcript variants encoding two different
isoforms have been found for this gene.
N-terminal 6His-tagged, recombinant, human MKP5, amino acids 320-end, expressed in E. coli.

Packaging

Also available in
250μg
size as catalog #
14-779M
and in
bulk
as catalog #
14-779-K
.

Quality

Routinely evaluated by the release of phosphate from the substrate 6,8-difluoro-4-methylumbelliferyl phosphate (DiFMUP)

Physical form

Protein in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.

Format: Purified using Ni2+/NTA agarose.

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service