Skip to Content
Merck
All Photos(1)

Documents

1.46613

Millipore

Sodium Chloride Peptone Broth

ready-to-use, bottle volume 250 mL , Filling volume

Synonym(s):

Nacl Broth, Peptone Broth, Sodium Chloride Peptone Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201

Agency

EP 2.6.13
JP 5.02
USP 62

Quality Level

form

liquid

feature

closure type screw cap with septum
ready-to-use

packaging

pack of 6 × 250 mL each
bottle of

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

250 mL

bottle volume

250 mL , Filling volume

pH range

6.8-7.2

application(s)

bioburden testing
pharmaceutical

storage temp.

2-25°C

suitability

nonselective for

General description

Sodium Chloride peptone broth is designed as a general diluent used for microbial examination of non-sterile pharmaceutical products. The low peptone concentration maintains viability without promoting growth. Presence of Sodium chloride maintains the osmotic balance of the medium and ensures cell integrity.
The neutral pH range is optimum for the viability of microorganisms′ present. The medium aims at maximum recovery of microorganisms present in the test sample.

Application

Sodium Chloride peptone broth is used as a protective and isotonic diluent for various microbiological test methods. The medium is used to maintain viability of the microorganisms present in the test sample.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service