Skip to Content
Merck
All Photos(2)

Key Documents

1.46285

Millipore

Tryptic Soy Agar - Ready-to-use Settle Plates

HEIMPLATE, Tween®, Histidine, Lecithin, Settle plate, irradiated, plate diam. 90 mm, suitable for air monitoring

Synonym(s):

CASO Agar, Casein Soybean Digest Agar, TSA, HEIMPLATE TSA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74
Pricing and availability is not currently available.

Quality Level

sterility

irradiated

form

plate (ready-to-use)

feature

ready-to-use

composition

Histidine
Lecithin
Tween®

packaging

pkg of (20 or 120 plates per box)

technique(s)

microbiological culture: suitable

plate diam.

90 mm

pH

7.3 ( in H2O)

application(s)

air monitoring
microbiology
pharmaceutical

storage temp.

2-8°C

suitability

aerobic bacteria (total count)

General description

Tryptic Soy Contact Agar + LTH is designed for the determination of the total aerobic microbial count on dry, sanitized surfaces and personnel in the non-critical area.
TSA medium is used for the cultivation and isolation of a wide range of bacteria, yeasts, and molds. The formulation contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. Sodium chloride maintains the osmotic equilibrium. The medium is also supplemented with neutralizers such as lecithin, polysorbate (Tween®) 80, and histidine to inactivate disinfectant residues.

Application

The TSA Settle plate is designed for the determination of the total aerobic microbial count on dry, sanitized surfaces, and personnel in the non-critical area.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service