Skip to Content
Merck
All Photos(1)

Documents

1.44244

Millipore

Hycon® Agar Strips SDX-y

Sabouraud Dextrose Agar, Irradiable antibiotics, for yeasts, for molds, γ-irradiated, double packed of 40 strips, suitable for air monitoring

Synonym(s):

Hycon Ready-to-use Agar Strips, SDX, SDA, Agar Strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113035
NACRES:
NA.74

Quality Level

sterility

γ-irradiated

form

plate (ready-to-use)

feature

individually wrapped

composition

Irradiable antibiotics

packaging

double packed of 40 strips

manufacturer/tradename

HYCON®

pH

6.9 ( in H2O)

application(s)

air monitoring
pharmaceutical

compatibility

for use with RCS® High Flow Touch

storage temp.

2-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Agar Strips SDX-y are ready-to-use culture media for assessment of airborne yeasts and molds with RCS® Air Samplers. Each individual agar strip is welded into a re-sealable sleeve and 100% visually controlled, ensuring maximum safety in critical environments. Sabouraud Dextrose Agar is filled in a flexible backing film. Sabouraud Dextrose Agar (SDX, SDA) is a complex medium for cultivation and isolation of a wide range of yeasts and molds. Dextrose supports the growth of fungi. The medium is supplemented with a mixture of irradiable antibiotics with inhibiting properties towards bacterial strains

Application

Agar Strips are used for monitoring of ambient air and compressed gases with RCS® Air Samplers. These Agar strips are gamma irradiated and triple bagged, for specific application within isolator and critical environment.

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany
RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

For easy and flexible microbiological monitoring of surfaces in the food & beverage industry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service