Skip to Content
Merck
All Photos(1)

Key Documents

1.44019

Millipore

Hycon® Contact Slides

MacConkey Agar, for enterobacteriaceae, sterile; aseptically filled, single packed of 20 units, suitable for surface monitoring

Synonym(s):

Hycon® Contact Slides C, MAC agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74
Pricing and availability is not currently available.

Quality Level

sterility

sterile; aseptically filled

form

plate (ready-to-use)

feature

ready-to-use

packaging

single packed of 20 units

manufacturer/tradename

HYCON®

pH

7.1 ( in H2O)

mp

>95 °C

application(s)

cosmetics
food and beverages
pharmaceutical

surface monitoring

compatibility

stand alone

storage temp.

2-25°C

suitability

enterobacteriaceae

General description

Contact Slides C are developed for easy and flexible microbiological monitoring of flat and curved surfaces. Each contact slide is individually sealed in the transparent and flexible primary backing film made from PET and provides a rectangular surface of 25 cm2. MacConkey Agar is a weak selective medium for the isolation of Enterobacteriaceae or specifically E. coli. Bile salts and crystal violet largely inhibit the growth of the gram-positive microbial flora, whereas the growth of most Gram-negative bacteria is supported. Lactose and the pH indicator neutral red are used to detect lactose degradation. Bacteria degrading lactose to acids grow in pink to red colored colonies. Additionally, Escherichia coli and other acid forming bacteria will show a zone of precipitated bile salt around the colonies. Bacteria not degrading Lactose will grow colorless.

Application

Contact Slides C are designed for the determination of the coliform and E. coli count on dry, sanitized surfaces as well as personnel in controlled clean room environments of production lines, i.e. food and beverages as well as non-sterile and terminally sterilized pharmaceuticals.

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service