Skip to Content
Merck
All Photos(1)

Key Documents

14-198

Sigma-Aldrich

MAP Kinase 2/Erk2 Protein, inactive, Mouse, 50 g

Unactive, recombinant full-length mouse p42 MAP Kinase 2/Erk2 fused with GST at the N terminus, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

recombinant

expressed in E. coli

mol wt

Mw 67.8 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Product Source: expressed in E coli
Recombinant full-length mouse p42 MAP Kinase 2/Erk2 fused with GST at the N terminus

Quality

routinely evaluated by phosphorylation of MBP (Catalog # 13-104) substrate in vitro.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tian-Rui Xu et al.
The Biochemical journal, 413(1), 51-60 (2008-04-26)
FRET (fluorescence resonance energy transfer) and co-immunoprecipitation studies confirmed the capacity of beta-arrestin 2 to self-associate. Amino acids potentially involved in direct protein-protein interaction were identified via combinations of spot-immobilized peptide arrays and mapping of surface exposure. Among potential key
Maofu Fu et al.
Methods in molecular biology (Clifton, N.J.), 284, 15-36 (2004-06-03)
Signal transduction pathways mediate cell-cell interactions and integrate signals from the extracellular environment through specific receptors at the cell membrane. They play a pivotal role in regulating cellular growth and differentiation and in mediating many physiological and pathological processes, such
Okano J et al.
The Journal of biological chemistry, 276(22), 19555-19564 (2001-03-30)
Paclitaxel is a widely used chemotherapeutic agent and is known to induce programmed cell death (apoptosis) in a variety of cell types, but the precise underlying mechanisms are poorly understood. To elucidate these mechanisms, we challenged human esophageal squamous cancer
D Aronson et al.
The Journal of biological chemistry, 272(41), 25636-25640 (1997-11-05)
Contractile activity plays a critical role in the regulation of gene transcription in skeletal muscle, which in turn determines muscle functional capabilities. However, little is known about the molecular signaling mechanisms that convert contractile activity into gene regulatory responses in
O Zugasti et al.
Molecular and cellular biology, 21(19), 6706-6717 (2001-09-05)
Signals from the extracellular matrix are essential for the survival of many cell types. Dominant-negative mutants of two members of Rho family GTPases, Rac1 and Cdc42, mimic the loss of anchorage in primary mouse fibroblasts and are potent inducers of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service