Skip to Content
Merck
All Photos(2)

Key Documents

14-1054

Sigma-Aldrich

Human recombinant histone H3.3-H4 tetramers

Human recombinant histone H3.3-H4 tetramers expressed in E Coli as fusion proteins with either an N-terminal FLAG or 6xHis epitopes & purified by Fast Protein Liquid Chromatography (FPLC).

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.32

biological source

Escherichia coli

recombinant

expressed in E. coli

Assay

>98% (SDS-PAGE)

manufacturer/tradename

Upstate®

concentration

1 μg/μL

technique(s)

activity assay: suitable

shipped in

dry ice

General description

Human recombinant histone H3.3-H4 tetramers expressed in E Coli. as fusion proteins with either an N-terminal FLAG® or 6xHis epitopes, and purified by Fast Protein Liquid Chromatography (FPLC).
Product Source: Human
The core histones, H2A, H2B, H3, and H4, form a central octameric structure that binds approximately 147 bp of DNA to form a nucleosome. The histone variant H3.3 replaces the conventional H3.1 histone in a wide range of nucleosomes in active genes, and are incorporated into chromatin independently of DNA synthesis. The structure of chromatin can be altered by post-translational modifications (PTM) occurring on the C- and N-terminal tails of core histones. Histone modifications can either provide or hinder access of key transcriptional complexes to DNA sequences and can regulate the expression of genes. These effects can be deciphered by applications such as chromatin immunoprecipiation (ChIP) which requires high quality chromatin and essential tools such as histone proteins and appropriate enzymes to assemble chromatin in vitro.

Application

Research Category
Epigenetics & Nuclear Function
Suitable for in vitro chromatin assembly and enzyme activity assays

Physical form

In buffer containing 20 mM Tris-Cl pH 7.9, 1 mM EDTA, 2 M NaCl, 0.5 mM PMSF and 1 mM DTT

Storage and Stability

Stable at -80ºC for 1 year from date of receipt.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service