Skip to Content
Merck
All Photos(1)

Documents

138000

Sigma-Aldrich

D-Amino Acid Oxidase Inhibitor

The D-Amino Acid Oxidase Inhibitor, also referenced under CAS 39793-31-2, controls the biological activity of D-Amino Acid Oxidase.

Synonym(s):

D-Amino Acid Oxidase Inhibitor, 4H-Thieno[3,2-b]pyrrole-5-carboxylic acid, D-AAO Inhibitor, Compound 8

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO2S
CAS Number:
Molecular Weight:
167.19
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

5% NaOH: 25 mg/mL
DMSO: 80 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C7H5NO2S/c9-7(10)5-3-6-4(8-5)1-2-11-6/h1-3,8H,(H,9,10)

InChI key

PMHDSACGRKBACK-UHFFFAOYSA-N

General description

A cell-permeable thienopyrrole compound that potently inhibits the cellular activity of transfected D-amino acid oxidase (DAO/DAAO/DAMOX/OXDA) in CHO cells (IC50 against human and rat DAO = 145 and 114 nM, respectively), while exhibiting no activity towards D-aspartate oxidase (DDO; IC50 >5 µM), P450 enzymes CYP3A4/2D6/3C9 (IC50 >10 µM), and a panel of over 150 other enzymes, receptors, and ion channels. Although in vivo treatment of Compound 8 (200 mg/kg, i.p.) is shown to effectively inhibit DAO activity in rat brain and kidney (by 96% and 80%, respectively), the resulting D-serine concentration increase in the periphery and central nervous system appears to be far from sufficient to produce adequate antipsychotic and cognitive enhancing effects.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Smith, S.M., et al. 2009. J. Pharmacol. Exp. Ther.328, 921.
Sparey, T., et al. 2008. Bioorg. Med. Chem. Lett.18, 3386.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service