Skip to Content
Merck
All Photos(1)

Key Documents

13-107

Sigma-Aldrich

Core Histones

Highly pure core histones proteins (H2A, H2B, H3 and H4) including H1 purified from chicken erythrocytes, suitable as a substrate for histone modification assays (HAT, HDAC, DNMT) and nucleosome assembly studies.

Synonym(s):

Core Histone Proteins

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

chicken

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (histone acetyltransferase (HAT))

shipped in

dry ice

General description

Histones are a family of basic proteins that associate with DNA in the nucleus and help condense it into chromatin. Some histones function as spools for the thread-like DNA to wrap around forming a nucleosome. Each nucleosome is made of DNA wrapped around eight histone proteins that function like a spool and are called a histone octamer. Each histone octamer is composed of two copies each of the histone proteins H2A, H2B, H3, and H4. The chain of nucleosomes is then wrapped into a 30 nm spiral called a solenoid, where additional H1 histone proteins are associated with each nucleosome to maintain the chromosome structure.

Application

Core histones and histone H1, purified from chicken erythrocytes; suitable as a substrate for HAT assays; purity = 90%

Quality

rountinely evaluated by immunoblot using Anti-Histone H3 (05-499) and Anti-Histone H4 (07-108)

Physical form

Lyophilized

Storage and Stability

Lyophilized: 2 years at -20°C; Rehydrated: 6 months at -70°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of nuclear receptor acetylation.
Chenguang Wang,Michael Powell,Lifeng Tian,Richard G Pestell
Methods in Molecular Biology null
1-Aminoindan-1,5-dicarboxylic acid and (S)-(+)-2-(3'-carboxybicyclo[1.1.1] pentyl)-glycine, two mGlu1 receptor-preferring antagonists, reduce neuronal death in in vitro and in vivo models of cerebral ischaemia.
D E Pellegrini-Giampietro,A Cozzi,F Peruginelli,P Leonardi,E Meli,R Pellicciari,F Moroni
The European Journal of Neuroscience null
Human erythropoietin-specific sites of monoclonal antibody-mediated neutralization.
M R Fibi,M Aslan,P Hintz-Obertreis,J U Pauly,M Gerken,G Luben,L Lauffer,B Siebold,W Stuber,G Nau
Blood null
Rocío Montes de Oca et al.
The Journal of biological chemistry, 280(51), 42252-42262 (2005-10-06)
Barrier to autointegration factor (BAF) is an essential conserved double-stranded DNA-binding protein in metazoans. BAF binds directly to LEM domain nuclear proteins (e.g. LAP2, Emerin, and MAN1), lamin A, homeodomain transcription factors, and human immunodeficiency virus type 1-encoded proteins. BAF
Christina Schellenbacher et al.
Journal of virology, 83(19), 10085-10095 (2009-07-31)
The amino (N) terminus of the human papillomavirus (HPV) minor capsid protein L2 can induce low-titer, cross-neutralizing antibodies. The aim of this study was to improve immunogenicity of L2 peptides by surface display on highly ordered, self-assembled virus-like particles (VLP)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service