1.25078
Fractogel® EMD TMAE Medcap (M)
MiniChrom Column Fractogel® TMAE Medcap (M)
Synonym(s):
MiniChrom Column Fractogel® TMAE Medcap (M), 5 ml
About This Item
Recommended Products
ligand
(Trimethylammoniumethyl)
material
polypropylene housing
product line
Fractogel®
parameter
≤8 bar pressure
~180 cm/hr max. flow rate
column I.D. × L
8 mm × 100 mm
volume
5 mL
matrix active group
Methacrylate
mean particle size
40-90 μm
capacity
~150 mg, BSA binding capacity
application(s)
plasma purification
recombinant protein
vaccine development
separation technique
anion exchange
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Excellent binding at high flow rates
- Excellent pressure-flow behavior
- Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution
Legal Information
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service