125059
Fractogel® EMD DMAE (M)
MiniChrom Fractogel® DMAE (M), volume 1 mL, prepacked column
Synonym(s):
MiniChrom Column Fractogel® DMAE (M), 1 ml
About This Item
Recommended Products
ligand
diethyl-(2-hydroxypropyl)aminoethyl
Quality Level
material
polypropylene housing
product line
Fractogel®
form
prepacked column
parameter
≤8 bar pressure
~170 cm/hr max. flow rate
column I.D. × L
8 mm × 20 mm
volume
1 mL
matrix active group
Methacrylate
mean particle size
40-90 μm
capacity
~100 mg, BSA binding capacity
application(s)
gene therapy
recombinant protein
vaccine development
shipped in
ambient
storage temp.
15-25°C
General description
Legal Information
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service