Skip to Content
Merck
All Photos(1)

Documents

12-340

Sigma-Aldrich

Akt/SGK Substrate Peptide

Akt/SGK Substrate Peptide primarily used in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.41

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

shipped in

dry ice

Biochem/physiol Actions

Protein Target: Akt
Target Sub-Family: AGC

Quality

Routinely evaluated by Kinase Assay as a substrate for Akt1/PKB and SGK active enzymes.

Physical form

sterile deionized water

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Guo et al.
The Journal of biological chemistry, 276(11), 8516-8523 (2000-12-16)
CAAT/enhancer-binding proteins (C/EBPs) play an important role in the regulation of gene expression in insulin-responsive tissues. We have found that a complex containing C/EBPbeta interacts with an insulin response sequence in the insulin-like growth factor-binding protein-1 (IGFBP-1) gene and that
D R Alessi et al.
FEBS letters, 399(3), 333-338 (1996-12-16)
The substrate specificity of protein kinase-B alpha (PKBalpha, also known as RAC kinase or Akt) was investigated using synthetic peptide substrates related to the sequence surrounding the phosphorylation site on glycogen synthase kinase-3 (GSK3). The minimum sequence motif required for
S Zhong et al.
The Journal of biological chemistry, 276(35), 33213-33219 (2001-07-07)
N-terminal tail phosphorylation of histone H3 plays an important role in gene expression, chromatin remodeling, and chromosome condensation. Phosphorylation of histone H3 at serine 10 was shown to be mediated by RSK2, mitogen- and stress-activated protein kinase-1 (MSK1), and mitogen-activated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service