Skip to Content
Merck
All Photos(1)

Key Documents

1.18394

Supelco

Phosphate Test Kit, colorimetric

0.015-0.14 mg/L (PO₄-P), 0.046-0.43 mg/L (PO₄³⁻), MQuant®

Synonym(s):

Phosphate Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Phosphate Test, colorimetric, 0.015-0.14 mg/L (PO4-P), 0.046-0.43 mg/L (PO43-), for use with MCOLORTEST®

specific analyte(s)

phosphate

Quality Level

measuring range

0.015-0.14 mg/L (PO4-P)
0.046-0.43 mg/L (PO43-)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric, PMB, with color card and comparator block with long test tubes 0.015 - 0.030 - 0.045 - 0.06 - 0.08 - 0.11 - 0.14 mg/l PO4-P 0.046 - 0.09 - 0.14 - 0.18 - 0.25 - 0.34 - 0.43 mg/l PO43- 0.034 - 0.07 - 0.10 - 0.14 - 0.18 - 0.25 - 0.32 mg/l P2O5 MQuant®
The MQuant® Phosphate test (formally MColortest®) can be used for the determination of orthophosphate in:
  • Groundwater and surface water, seawater
  • Drinking water
  • Wastewater
  • Soils and fertilizers after appropriate sample pretreatment
  • Food after appropriate sample pretreatment
  • Steels after appropriate sample pretreatment

Method
In sulfuric solution orthophosphate ions react with molybdate ions to form molybdophosphoric acid. Ascorbic acid reduces this to phosphomolybdenum blue (PMB). The phosphate concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card. The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range.
Due to the long test tubes a sensitive measurement range can be covered.
Using the refill pack 1.18465 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.

Application


  • Environmental Monitoring: Phosphate tests are extensively used in environmental monitoring to assess the levels of phosphate in water bodies. These tests help in evaluating the impact of agricultural runoff, industrial discharge, and wastewater treatment on water quality. Recent studies have focused on using selective colorimetric nano-sensing solutions to determine phosphate ions in drinking water samples, highlighting their importance in ensuring safe and clean water supplies (Salem and Draz, International Journal of Environmental Analytical Chemistry, 2020).

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service