Skip to Content
Merck
All Photos(1)

Documents

1.18303

Supelco

Butane-1-sulfonic acid sodium salt

for ion pair chromatography LiChropur

Synonym(s):

Butane-1-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NaO3S
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
12352107
EC Index Number:
219-201-1
NACRES:
NA.21

Quality Level

Assay

≥99% (acidimetric)

technique(s)

ion pair chromatography: suitable

transmittance

200 nm, ≥70%
220 nm, ≥90%
250 nm, ≥98%
(0.005 M; 1 cm in H2O)

pH

5.0-7.0 (100 g/L in H2O)

mp

>310 °C

storage temp.

15-25°C

InChI

1S/C4H10O3S.Na/c1-2-3-4-8(5,6)7;/h2-4H2,1H3,(H,5,6,7);/q;+1/p-1

InChI key

XQCHMGAOAWZUPI-UHFFFAOYSA-M

General description

Butane-1-sulfonic acid sodium salt is commonly used as an ion-pairing reagent in ion-pair chromatography (IPC).

Application

Butane-1-sulfonic acid sodium salt can be used as anion-pair agent in the determination of biogenic amines (BA):

  • In tomato by ion-pair chromatography coupled toa potentiometric detector.
  • Inalcoholic beverages by HPLC coupled to a potentiometric detector for foodquality control.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

HPLC-potentiometric method for determination of biogenic amines in alcoholic beverages: A reliable approach for food quality control
Gil RL, et al.
Food Chemistry, 372, 131288-131288 (2022)
Determination of biogenic amines in tomato by ion-pair chromatography coupled to an amine-selective potentiometric detector
Gil R, et al.
Electrochimica Acta, 378, 138134-138134 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service