Skip to Content
Merck
All Photos(1)

Documents

1.14783

Supelco

Nickel Test Kit, colorimetric

0.5-10 mg/L (Ni), MQuant®

Synonym(s):

Nickel Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Nickel Test, colorimetric, 0.5-10 mg/L (Ni), for use with MQUANT®

specific analyte(s)

nickel

Quality Level

measuring range

0.5-10 mg/L (Ni)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color-disk comparator 0.5 - 1.0 - 1.5 - 2 - 3 - 4 - 6 - 8 - 10 mg/l Ni MQuant®
The MQuant® Nickel test (formally MColortest®) measures nickel(II) ions in
  • Groundwater and surface water
  • Drinking water
  • Industrial water
  • Wastewater and percolating water
  • Electroplating wastewater
This test is not suited for seawater.

Method
Nickel(II) ions are oxidized by iodine and then transformed with dimethylglyoxime in an ammoniacal solution into a red-brown complex. The nickel concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation.
The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.
Using the refill pack 1.18461 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.
The test measures nickel(II) ions in groundwater and surface water, drinking water, industrial water and wastewater.

Analysis Note

Application testconforms

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service