Skip to Content
Merck
All Photos(1)

Key Documents

114666

Sigma-Aldrich

Actinomycin D, Streptomyces sp.

Anti-neoplastic antibiotic. Inhibits DNA-primed RNA polymerase by complexing with DNA via deoxyguanosine residues.

Synonym(s):

Actinomycin D, Streptomyces sp., Dactinomycin, RNA Polymerase I Inhibitor I, Pol I Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H86N12O16
CAS Number:
Molecular Weight:
1255.42
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

red

solubility

DMSO: 1 mg/mL
chloroform: soluble
methanol: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C62H86N12O16/c1-27(2)42-59(84)73-23-17-19-36(73)57(82)69(13)25-38(75)71(15)48(29(5)6)61(86)88-33(11)44(55(80)65-42)67-53(78)35-22-21-31(9)51-46(35)64-47-40(41(63)50(77)32(10)52(47)90-51)54(79)68-45-34(12)89-62(87)49(30(7)8)72(16)39(76)26-70(14)58(83)37-20-18-24-74(37)60(85)43(28(3)4)66-56(45)81/h21-22,27-30,33-34,36-37,42-45,48-49H,17-20,23-26,63H2,1-16H3,(H,65,80)(H,66,81)(H,67,78)(H,68,79)

InChI key

RJURFGZVJUQBHK-UHFFFAOYSA-N

General description

Anti-neoplastic antibiotic. Inhibits DNA-primed RNA polymerase by complexing with DNA via deoxyguanosine residues. At higher concentrations, DNA polymerase is inhibited. Also acts as a competitive inhibitor of serine proteases. Inhibits cell growth and colony formation in synchronized HeLa cells. Induces apoptosis in many cell lines. However, actinomycin D has also been shown to suppress etoposide-induced apoptosis in PC12 cells. Note: 1 set = 20 x 200 µg.
Antineoplastic antibiotic. Inhibits DNA-primed RNA polymerase by complexing with DNA via deoxyguanosine residues. A DNA-intercalator with growth inhibitory activity against certain leukemias and sarcomas. At higher concentrations, DNA polymerase is inhibited. Inhibits cell growth and colony formation in synchronized HeLa cells. A potent inducer of apoptosis in many cell lines; however, actinomycin D has also been shown to suppress programmed cell death of PC12 cells induced by etoposide, an inhibitor of topoisomerase II. Acts as a competitive inhibitor of serine protease from microorganisms. The growth of S. aureus is inhibited by 0.25-0.75 µg/ml, whereas a concentration of 100 µg/ml is required to inhibit E. coli due to permeability differences. Heat resistant and stable between pH 2 and 11; decomposed by strong acids and bases. λmax 441 nm ±2 nm. Note: 1 set = 20 vials x 200 µg.
Stability: heat resistant and stable between pH 2 and 11. Decomposed by strong acids and bases.

Biochem/physiol Actions

Primary Target
serine proteases
Secondary Target
cell growth and colony formation in synchronized HeLa cells

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Nakajima, M., et al. 1994. Neurosci. Lett.176, 161.
Wu, M.H., and Yung, B.Y. 1994. Eur. J. Pharmacol. 270, 203.
Betzel, C., et al. 1993. Biochim. Biophys. Acta 1161, 47.
Yung, B.Y., et al. 1992. Int. J. Cancer52, 317.
Martin, S.J., et al. 1990. J. Immunol.145, 1859.
Yung, B.Y., et al. 1990. Cancer Res.50, 5987.
White, R.J., and Phillips, D.R. 1985. Biochemistry27, 9122.
Madharavao, M., et al. 1978. J. Med. Chem.21, 958.
Sengupta, S.K., et al. 1975. J. Med. Chem.18, 1175.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Azam Asgarihafshejani et al.
iScience, 25(5), 104259-104259 (2022-05-07)
Hippocampal somatostatin (SOM) cells are dendrite-projecting inhibitory interneurons. CA1 SOM cells receive major excitatory inputs from pyramidal cells (PC-SOM synapses) which show mGluR1a- and mTORC1-mediated long-term potentiation (LTP). PC-SOM synapse LTP contributes to CA1 network metaplasticity and memory consolidation, but
Sarah V Schweighofer et al.
Cell death and differentiation, 31(4), 469-478 (2024-03-20)
One hallmark of apoptosis is the oligomerization of BAX and BAK to form a pore in the mitochondrial outer membrane, which mediates the release of pro-apoptotic intermembrane space proteins into the cytosol. Cells overexpressing BAX or BAK fusion proteins are
Leonid Anikin et al.
International journal of molecular sciences, 23(3) (2022-02-16)
Aminoacridines, used for decades as antiseptic and antiparasitic agents, are prospective candidates for therapeutic repurposing and new drug development. Although the mechanisms behind their biological effects are not fully elucidated, they are most often attributed to the acridines' ability to
Yu Zhou et al.
iScience, 27(2), 108822-108822 (2024-02-02)
Alternative polyadenylation (APA) is an important post-transcriptional regulatory mechanism and is involved in many diseases, but its function and mechanism in regulating pancreatic cancer (PC) pathogenesis remain unclear. In this study, we found that the 3' UTR shortening of MZT1
Viana Manrique-Suárez et al.
Proteins, 89(11), 1508-1521 (2021-07-06)
Tumor necrosis factor-alpha (TNFα) inhibitors could prevent neurological disorders systemically, but their design generally relies on molecules unable to cross the blood-brain barrier (BBB). This research was aimed to design and characterize a novel TNFα inhibitor based on the angiopeptide-2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service