Skip to Content
Merck
All Photos(1)

Documents

1.13720

Sigma-Aldrich

Dichloromethane-D2

deuteration degree min. 99.8% for NMR spectroscopy MagniSolv

Synonym(s):

Dichloromethane-D2, Dideuteromethylene chloride, Methylene chloride-d2, Dideuterodichloromethane, DCM deuterated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CCl2D2
CAS Number:
Molecular Weight:
86.94
MDL number:
UNSPSC Code:
12142201
EC Index Number:
216-776-0
NACRES:
NA.22

vapor pressure

450 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

605 °C (DIN 51794)

potency

1600 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

13-22 % (v/v)

pH

7 (20 °C)

bp

39 °C/1013 hPa

mp

-97 °C

density

1.36 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/CH2Cl2/c2-1-3/h1H2/i1D2

InChI key

YMWUJEATGCHHMB-DICFDUPASA-N

General description

A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.

Analysis Note

Deuteration degree: ≥ 99.8 %
Water (NMR, H₂O): ≤ 0.01 %
Performance test (NMR-spectrum): passes test

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service