Skip to Content
Merck
All Photos(1)

Documents

1.09881

Supelco

Buffer concentrate

(glycine/hydrochloric acid), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 1.00 ± 0.02 (20°C) Titrisol®

Synonym(s):

Titrisols for preparation of buffer solutions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161704
NACRES:
NA.25

grade

analytical standard

Quality Level

Agency

according to DIN 19266

product line

Titrisol®

pH

1.00 (20 °C in H2O)

density

1.02 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The Titrisol® reference buffer concentrate [glycine/hydrochloric acid; pH=1.00±0.02 (20 °C)] is analyzed in the ISO/IEC 17025 accredited calibration laboratory with a combined glass electrode and 5-point calibration according to DIN 19268 against a certified primary standard reference material following DIN 19266.

The Titrisol® concentrated buffer material combines ease of use and maximum precision for the routine calibration and monitoring of pH instruments. To prepare the ready-to-use pH buffer solution, simply dilute the Titrisol® buffer concentrate.

Features and Benefits

  • Traceable to primary standard reference materials from NIST and PTB
  • Analytical standard Grade
  • Uncertainty data and traceability are documented in a comprehensive, batch-specific Certificate of Analysis.

Analysis Note

pH-value (20 °C): 1.00 +/- 0.02
Measured after dilution to 500 ml with a combination glass electrode after multiple point calibration (according to DIN 19268) with reference materials for the preparation of standard buffer solutions according to DIN 19266.

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

standard

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service