Skip to Content
Merck
All Photos(1)

Key Documents

1.09752

Supelco

Photometer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115311

technique(s)

photometry: suitable

Quality Level

compatibility

for use with Spectroquant® Nova 60 A

storage temp.

2-30°C

General description

Enjoy high measurement quality with great simplicity. The Spectroquant NOVA 60 A packs maximum convenience into a minimum size. With an integrated (recharge) battery pack and its robust design this compact filter photometer follows you wherever it is needed.

The Spectroquant NOVA 60 A uses 12 filters in array-technique to cover the most important analytical wavelength to be used with our Spectroquant Test Kits. Moreover, the integrated bar-code reading of Spectroquant tests for auto-recognition of cell size, method parameters, and calculation of results guarantees reproducible and accurate results are produced even without excessive training.
NOVA 60 A Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A step-by-step procedure for the photometric determination of Nitrate in effluents using Spectroquant® test kits and photometer for a precise and quantitative analysis.

Protocols

Step-by-step method describing the photometric determination of ammonium ions in effluents using Spectroquant® test kits and spectrophotometer.

Spectrophotometric determination of bromine with dipropyl-p-phenylenediamine (DPD) in drinking water, swimming-pool water, wastewater, and disinfection solutions in a range of 0.020 – 10.00 mg/L.

Cobalt in wastewater from the production and casting of non-ferrous metals

Related Content

Learn about the sample preparation and photometric determination of non-ionic surfactants in Extran® rinse solutions (MA 01, MA 02, MA 05, AP 22) using Spectroquant® test kits and spectrophotometers.

Questions

  1. Can this analyzer measure turbidity in liquid samples?

    1 answer
    1. The Nova 60 A is discontinued. It may be worth considering a turbidimeter for turbidity measurements such as items 1.18324 or 1.18325.

      https://www.sigmaaldrich.com/search/turbidimeter?focus=products&page=1&perpage=30&sort=relevance&term=turbidimeter&type=product

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service