Skip to Content
Merck
All Photos(1)

Key Documents

1.09748

Supelco

Photometer

for use with SPECTROQUANT® NOVA 30 A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115311

compatibility

for use with SPECTROQUANT® NOVA 30 A

storage temp.

2-30°C

General description

NOVA 30 A Spectroquant®
The Spectroquant NOVA 30 A photometer is the basic instrument for comfortable wastewater analysis. It is compact, robust, and - whenever the circumstances demand - mobile, too. Additionally this basic model incorporates advanced features like barcode reading for all tests and instrument-supported AQA (Analytical Quality Assurance).

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service