Skip to Content
Merck
All Photos(1)

Documents

1.09570

Supelco

pH-indicator paper pH 9.5 - 13.0

roll L 4.8 m, pkg of 6 rolls, Alkalit

Synonym(s):

pH chemical test paper paper pH 9.5 - 13.0, pH test paper paper pH 9.5 - 13.0

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113034

feature

non-bleeding: no

Quality Level

packaging

pkg of 6 rolls

manufacturer/tradename

Alkalit

roll L

4.8 m

detection method

colorimetric

storage temp.

15-25°C

General description

6 replacement rolls (of 4.8 m), for 109562, 109565 Alkalit

MQuant® pH indicator papers supplied in rolls are perfectly suited for longterm storage, since they are protected against external influences, such as moisture, light, and ambient gases. These pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. The paper is always ready to use; it is easily pulled out of the plastic reel and can be torn off in the necessary length. The pH value is measured semiquantitatively by visual comparison of the color formed on the paper with the fields of the color scale. This article contains replacement rolls without color scale for article 109562.

Legal Information

ALKALIT is a trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service