Skip to Content
Merck
All Photos(2)

Documents

1.09489

Supelco

pH-indicator paper Litmus paper red

roll L 4.8 m , with color scale, reag. Ph. Eur., MQuant®

Synonym(s):

pH chemical test paper paper Litmus paper red, pH test paper paper Litmus paper red

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NB.35

Agency

reag. Ph. Eur.

Quality Level

product line

MQuant®

feature

non-bleeding: no

roll L

4.8 m , with color scale

detection method

colorimetric

storage temp.

15-25°C

General description

MQuant® Litmus paper red is suitable to monitor a pH change from acidic to alkalinic. The paper changes its color from red (below 4.5) to blue (above pH 8.3). Litmus is a natural product; it is the oldest form of pH indicator absorbed on paper. This is why litmus paper is still broadly used as a synonyme for other pH papers, eventhough these are impregnated with other, mostly synthetic, indicator dyes. pH indicator papers supplied in rolls are better suited for longterm storage, since they are protected against external influences. For a semi-quantitative monitoring of the pH value, please refer to our MQuant® pH-indicator papers or strips and for a quantitative measurement to our Reflectoquant® test strips.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service