Skip to Content
Merck
All Photos(1)

Key Documents

1.08426

Supelco

Titriplex® V

for analysis (diethylenetriaminepentaacetic acid)

Synonym(s):

Titriplex® V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23N3O10
CAS Number:
Molecular Weight:
393.35
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-652-8
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: Complexometric reactions

technique(s)

titration: suitable

pH

2-3 (20 °C in H2O, saturated solution)

mp

>206 °C (Decomposition)

solubility

4.8 g/L

bulk density

580 kg/m3

storage temp.

15-25°C

SMILES string

N(CCN(CC(=O)O)CC(=O)O)(CCN(CC(=O)O)CC(=O)O)CC(=O)O

InChI

1S/C14H23N3O10/c18-10(19)5-15(1-3-16(6-11(20)21)7-12(22)23)2-4-17(8-13(24)25)9-14(26)27/h1-9H2,(H,18,19)(H,20,21)(H,22,23)(H,24,25)(H,26,27)

InChI key

QPCDCPDFJACHGM-UHFFFAOYSA-N

Analysis Note

Assay (complexometric, calc. on anhydrous substance): ≥ 99 %
Identity (IR-spectrum): passes test
Heavy metals (as Pb): ≤ 0.001 %
Iron (Fe): ≤ 0.001 %
Sulfated ash: ≤ 0.1
Water (according to Karl Fischer): ≤ 0.5

Legal Information

Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

392.0 °F - closed cup

Flash Point(C)

200 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service