Skip to Content
Merck
All Photos(1)

Documents

1.08424

Supelco

Titriplex® IV

suitable for titration

Synonym(s):

Titriplex® IV, 1,2-Cyclohexanediamine-N,N,N′N′-tetraacetic acid, 1,2-Cyclohexylenedinitrilotetraacetic acid, 1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O8 · xH2O
CAS Number:
Molecular Weight:
346.33 (anhydrous basis)
MDL number:
UNSPSC Code:
12352106
EC Index Number:
236-308-9
NACRES:
NA.21

product name

Titriplex® IV, for analysis (1,2-cyclohexylenedinitrilotetraacetic acid monohydrate)

Quality Level

form

solid

reaction suitability

reaction type: Complexometric reactions

technique(s)

titration: suitable

mp

213 °C

bulk density

500 kg/m3

storage temp.

no temp limit

InChI

1S/C14H22N2O8/c17-11(18)5-15(6-12(19)20)9-3-1-2-4-10(9)16(7-13(21)22)8-14(23)24/h9-10H,1-8H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24)/p-2/t9-,10-/m1/s1

InChI key

FCKYPQBAHLOOJQ-NXEZZACHSA-L

Analysis Note

Assay (complexometric, calc. on anhydrous substance): ≥ 99.0 %
Identity (IR-spectrum): passes test
Sulphate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Sulfated ash: ≤ 0.1 %
Water (according to Karl Fischer): 4.8 - 5.8 %

Legal Information

Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service