Skip to Content
Merck
All Photos(1)

Key Documents

1.05874

Supelco

Magnesium perchlorate hydrate

[about 83% Mg(ClO₄)₂] for analysis EMSURE®

Synonym(s):

Magnesium perchlorate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(ClO4)2 · x H2O
CAS Number:
Molecular Weight:
223.21 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-108-3
NACRES:
NA.21
form:
solid
solubility:
500 g/L
Pricing and availability is not currently available.

Quality Level

product line

EMSURE®

Assay

≥83% (complexometric)

form

solid

impurities

≤0.05% Free acid (as perchloric acid)
≤0.05% Free alkali (as magnesium oxide)

pH

8.2 (25 °C, 50 g/L in H2O)

solubility

500 g/L

density

2.60 g/cm3 at 25 °C (anhydrous substance)

bulk density

600‑800 kg/m3

anion traces

chloride (Cl-): ≤0.005%
nitrate (NO3-): ≤0.002%

storage temp.

2-30°C

SMILES string

[Mg+2].[Cl](=O)(=O)(=O)[O-].[Cl](=O)(=O)(=O)[O-].O

InChI

1S/2ClHO4.Mg.H2O/c2*2-1(3,4)5;;/h2*(H,2,3,4,5);;1H2/q;;+2;/p-2

InChI key

KKFFNQXCJMNXHI-UHFFFAOYSA-L

Related Categories

Linkage

Replaces: MX0069A-1

Analysis Note

Assay (complexometric): ≥ 83 %
Chloride (Cl): ≤ 0.005 %
Nitrate (NO₃): ≤ 0.002 %
Free alkali (as magnesium oxide): ≤ 0.05 %
Free acid (as perchloric acid): ≤ 0.05 %
Particle size (1-4 mm): about 75%

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service