Skip to Content
Merck
All Photos(1)

Documents

1.04660

Supelco

UV-VIS Standard 1a: Potassium dichromate solution (600 mg/l)

according to Ph. Eur. absorbance at 430nm, Certipur®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

according to Ph. Eur. absorbance at 430nm

product line

Certipur®

technique(s)

UV/Vis spectroscopy: suitable

application(s)

environmental
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

Easily meet your UV-VIS performance check requirements with an all-in-one qualification kit
for the absorbance at 430 nm according to Ph Eur Certipur®

Application

For additional information, please click Instructions for Use

Features and Benefits

  • It is a solution of potassium dichromate in sulfuric acid
  • Traceability to standard reference material from the National Institute of Standards and Technology (NIST)
  • Includes a certificate of analysis (CoA)
  • Reliable and reproducible results

Packaging

Each package includes 2x10 mL potassium dichromate solution and 6x10 mL sulfuric acid solution (used for baseline correction).
Each package will provide enough material for two complete instrument qualifications: including cell conditioning, baseline correction, standard measurement and baseline verification.

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service