Skip to Content
Merck
All Photos(1)

Documents

1.04413

Supelco

Mercury(II) sulfate solution 200 g/l

in diluted sulfuric acid for determination of COD acc. to DIN 38409-H 43-1 (short-term experience)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
SE.11

Quality Level

form

liquid

analyte functional class(es)

chemical oxygen demand (COD)

technique(s)

photometry: suitable

pH

0.5 (20 °C in H2O)

density

1.3 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

Concentration (HgSO₄): 198.0 - 202.0 g/l
Concentration (H₂SO₄): 173.0 - 180.0 g/l

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service