Skip to Content
Merck
All Photos(1)

Key Documents

1.03059

Supelco

5-(4-Dimethylaminobenzylidene)-rhodanine

GR for analysis (reagent for silver)

Synonym(s):

5-(4-Dimethylaminobenzylidene)-rhodanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2OS2
CAS Number:
Molecular Weight:
264.37
MDL number:
UNSPSC Code:
12352115
EC Index Number:
208-625-2
NACRES:
NA.05

Quality Level

Assay

≥98% (GC)

form

solid

mp

275-280 °C (decomposes)

bulk density

225 kg/m3

storage temp.

2-30°C

InChI

1S/C12H12N2OS2/c1-14(2)9-5-3-8(4-6-9)7-10-11(15)13-12(16)17-10/h3-7H,1-2H3,(H,13,15,16)/b10-7-

InChI key

JJRVRELEASDUMY-YFHOEESVSA-N

Analysis Note

Assay (of S): ≥ 98.0 %
Identity (IR): passes test
Substance insoluble in Aceton: passes test
Suitability for silver determination: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service