1.02747
Linden Grain Medium Halal (Base)
Suitable for use in Halal and Kosher certified processes, suitable for sterility testing
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
solid
application(s)
food and beverages
microbiology
sterility testing
storage temp.
2-8°C
suitability
bacteria
molds
yeasts
Related Categories
General description
Linden Grain Medium Halal (Base) is a powdered dehydrated culture media with high nutrient content. It is used for microbiological validation of aseptic filling lines across the beverage industry because it creates optimum growth conditions for fungi and bacteria at various pH levels. The pH of the medium can be adjusted to the filling line criteria. At a pH of 4.3, fungi and bacteria which are adapted to grow under high acid conditions show visible growth within 7 days of incubation at 28°C. Bacteria that grow best at neutral to slightly alkaline pH, like Bacillus subtilis, show no growth/very limited growth at pH 4.3. Any growth within the medium (turbidity), on the surface of the medium, or the inner wall or bottom of the tubes and bottles indicates the presence of spoilage organisms. Experiments have shown that the medium could also be used after freezing and thawing, thus making this medium suitable for ice crusher installations. This medium is halal-certified.
Application
Linden Grain Medium Halal (Base) is suitable for the detection of yeasts, molds, and bacteria in beverages. It is designed for media fill simulations of beverage bottling and ice crusher installations.
Footnote
We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Halal-certified culture media prevent non-halal ingredient contamination, enabling microbiological QC testing without losing certification.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service