Skip to Content
Merck

1.02485

SAFC

Cellvento® CHO-210

≤1.0 IU/mL endotoxin, pH 3.9-4.5(without supplement)

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
Pricing and availability is not currently available.

Quality Level

description

shipped with cooling packs

product line

Cellvento®

form

dry powder

technique(s)

cell culture | mammalian: suitable (suspension)

storage temp.

2-8°C

General description

Cellvento® CHO-210 medium has been specially developed for the growth of Chinese Hamster Ovary (CHO) cells and the expression of monoclonal antibodies and recombinant proteins in suspension culture for use in fed-batch manufacturing processes. The formulation is of non-animal origin, chemically defined and contains no hydrolysates or components of unknown composition. Cellvento® CHO-210 medium has been formulated without L-glutamine, hypoxanthine, thymidine, or thymidine, to keep flexibility in applications. It is available in dry powder from or as ready-to-use medium to fit to different experimental set-ups.

Application

Cellvento® CHO-210 medium has been designed to support optimal cell growth and performance of DHFR-negative CHO suspension cell types, primarily recombinant CHO-DG44 cell lines, but may be suitable for use with other CHO cell lines. It should be used as a production medium starting with the final N-1 expansion step in fed-batch applications, and then together with its companion feed product Cellvento® Feed-210 during the final production culture. This product is intended for research or further manufacturing but not for human or therapeutic use.

Quantity

Formulated to contain 23.13 grams of powder per liter of medium.

Legal Information

CELLVENTO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service