Skip to Content
Merck
All Photos(1)

Documents

1.01182

Supelco

Ammonium heptamolybdate tetrahydrate

GR for analysis ACS,ISO,Reag. Ph Eur

Synonym(s):

Ammonium heptamolybdate tetrahydrate, Ammonium molybdate, Hexammonium heptamolybdate 4-hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)6Mo7O24 · 4 H2O
CAS Number:
Molecular Weight:
1235.86
MDL number:
UNSPSC Code:
12352302
EC Index Number:
234-722-4
NACRES:
NB.21

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

form

solid

specific analyte(s)

phosphate

technique(s)

photometry: suitable

pH

5.3 (20 °C, 50 g/L in H2O)

mp

90 °C (Elimination of water of crystallisation)

solubility

400 g/L

density

2.498 g/cm3 at 20 °C

bulk density

800 kg/m3

storage temp.

2-30°C

InChI

1S/Mo.H3N.H2O.O/h;1H3;1H2;/q;;;-2/p+1

InChI key

XCOMAVCHADVANQ-UHFFFAOYSA-O

Looking for similar products? Visit Product Comparison Guide

Application


  • A Novel Hydro-Thermal Synthesis of Nano-Structured Molybdenum-Iron Intermetallic Alloys at Relatively Low Temperatures.: This study outlines a novel hydro-thermal method for synthesizing molybdenum-iron intermetallic alloys using lower temperatures, highlighting a potential route for cost-effective and energy-efficient material production (El-Geassy et al., 2023).

  • Alternative Preparation of Improved NiMo-Alumina Deoxygenation Catalysts.: This research discusses an alternative method for preparing NiMo-alumina catalysts with enhanced deoxygenation properties, potentially useful for synthetic and petrochemical applications (Priecel et al., 2020).

  • Chromatographic identification of "green capping agents" extracted from Nasturtium officinale (Brassicaceae) leaves for the synthesis of MoO(3) nanoparticles.: The paper identifies plant-based capping agents for synthesizing molybdenum trioxide nanoparticles, contributing to greener nanotechnology practices (Shaheen & Ahmad, 2020).

  • Preparation of h-MoO3 and α-MoO3 nanocrystals: comparative study on photocatalytic degradation of methylene blue under visible light irradiation.: This study compares the efficiency of hexagonal and orthorhombic molybdenum trioxide nanocrystals in degrading methylene blue, offering insights into their photocatalytic behaviors (Chithambararaj et al., 2013).

  • Novel MoO2/carbon hierarchical nano/microcomposites: synthesis, characterization, solid state transformations and thiophene HDS activity.: The research introduces a new type of molybdenum dioxide-carbon composite material with applications in hydrodesulfurization, demonstrating significant industrial potential (Avendaño et al., 2013).

Analysis Note

Assay (as MoO₃): 81.0 - 83.0 %
Assay ((NH₄)₆Mo₇O₂₄ x 4 H₂O): ≥ 99.0 %
Insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.0005 %
Nitrate (NO₃): passes test
Phosphate (PO₄): ≤ 0.0005 %
Phosphate, Arsenate, Silicate (as PO₄): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (ACS): ≤ 0.001 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.005 %
K (Potassium): ≤ 0.002 %
Na (Sodium): ≤ 0.01 %
Pb (Lead): ≤ 0.001 %
Corresponds to ACS,ISO,Reag. Ph Eur

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service