Skip to Content
Merck
All Photos(1)

Documents

1.00929

Supelco

Diethyl ether

≥99.9% (GC), contains 5-10 ppm 2, 6-Di-tert-butyl-4-methylphenol (BHT) as stabilizer

Synonym(s):

Et2O, Ethoxyethane, Ethyl ether, Ether, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
MDL number:
UNSPSC Code:
12191502

product name

Diethyl ether,

vapor density

2.6 (vs air)

vapor pressure

587 hPa ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

320 °F

potency

1215 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

contains

5-10 ppm 2,6-Di-tert-butyl-4-methylphenol (BHT) as stabilizer

expl. lim.

36.5 %

impurities

≤0.00003% Peroxide (as H2O2)
≤0.0002 meq/g Alkalinity
≤0.0005 meq/g Acidity
≤0.001% Aldehydes (as CH3CHO)
≤0.001% Carbonyl compounds (as CO)
≤0.005% Acetone (GC)
≤0.02% Ethanol (GC)
≤0.02% Methanol (GC)

evapn. residue

≤10 mg/L

color

APHA: ≤10

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

transition temp

flash point -40 °C

density

0.706 g/mL at 25 °C (lit.)

storage temp.

2-25°C

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SeccoSolv ready-to-use dried solvents fulfill even the most stringent requirements. They are produced using specially selected distillation methods that ensure consistently high dryness and batch-to-batch consistency. To protect the quality of these products even better from potential contaminants, our new SeccoSept septum seal cap provides multiple layers of protection to keep solvents in flawless condition before, during, and after removal.

Analysis Note

Purity (GC): ≥ 99.9 %
Identity (IR): conforms
Color: ≤ 10 Hazen
Acidity: ≤ 0.0005 meq/g
Alkalinity: ≤ 0.0002 meq/g
Acetone (GC): ≤ 0.005 %
Ethanol (GC): ≤ 0.02 %
Methanol (GC): ≤ 0.02 %
Aldehydes (as CH₃CHO): ≤ 0.001 %
Carbonyl compounds (as CO): ≤ 0.001 %
Peroxide (as H₂O₂): ≤ 0.00003 %
Evaporation residue: ≤ 10 mg/l
Water: ≤ 0.005 %
The product is stabilized with 5 - 10 ppm 2,6-Di-tert-butyl-4-methylphenol (BHT).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service