Skip to Content
Merck
All Photos(1)

Documents

1.00680

Supelco

AOX Standard

8-16 quality tests 0.2 - 2.0 mg/l Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Quality Level

analyte functional class(es)

adsorbable organic halides (AOX)

technique(s)

photometry: suitable

pH

5 (20 °C in H2O)

density

1.00 g/cm3 at 20 °C

format

single component solution

storage temp.

15-25°C

General description

The Spectroquant® AOX Standard is diluted with distilled water to prepare a standard solution of an appropriate concentration. This standard solution is then analyzed using the Spectroquant® AOX Cell Test to check the measurement procedure. AOX Standard takes the form of an aqueous solution of 4-chlorophenol with a concentration of 20 mg/l AOX.

These Spectroquant® standards perfectly complement our ready-to-use test kits and photometers within the Spectroquant portfolio and are ideal for validating international norm methods described in e.g., ISO, EN, APHA, and EPA.

Please visit the Reference Material Quality Grade website to find a comprehensive overview of our different grades of standards.®

Application

AOX Standard can be used to check the photometric measurement system (reagents, measurement device, handling) and the mode of working.

Features and Benefits

  • The standard is analogous to DIN EN ISO 9562
  • Accompanied with a certificate of analysis (COA)
  • Reliable and reproducible results

Analysis Note

Assay (AOX): 19.5 - 20.5 mg/l

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service