Skip to Content
Merck
All Photos(2)

Key Documents

07-849-I

Sigma-Aldrich

Anti-phospho-JAK1 (Tyr1022/1023)

from rabbit, purified by affinity chromatography

Synonym(s):

Tyrosine-protein kinase JAK1, Tyr1022/1023-phosphorylated, Janus kinase 1, Tyr1022/1023-phosphorylated, JAK-1, Tyr1022/1023-phosphorylated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), mouse (based on 100% sequence homology), canine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), porcine (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pTyr1022/pTyr1023)

Gene Information

human ... JAK1(3716)

General description

Tyrosine-protein kinase JAK1 (EC 2.7.10.2; UniProt P23458; also known as Janus kinase 1, JAK-1) is encoded by the JAK1 (also known as JAK1A, JAK1B) gene (Gene ID 3716) in human. JAK1 belongs to the family of non-receptor Janus tyrosine kinases (JAKs) that regulate a spectrum of cellular functions downstream of activated cytokine receptors in the lympho-hematopoietic system. Immunological stimuli, such as interferons and cytokines, cause the recruitment of Stat transcription factors to cytokine receptor. Subsequent phosphorylation by receptor-associated JAK causes Stat dimerization, nuclear translocation, and Stat-mediated target genes transcription. The canonical JAK/Stat pathway is integral to maintaining a normal immune system by stimulating proliferation, differentiation, survival, and host resistance to pathogens. Altering JAK/Stat signaling to reduce immune cells cytokine responses represents an attractive target for anti-inflammatory therapies. Although JAK1 is a ubiquitously expressed Janus kinase activated by a wide variety of cytokines, studies conducted on Jak1-knockout mice and cells indicate that JAK1 plays a non-redundant role in promoting biologic responses induced selectively by a subset of cytokine receptors.

Specificity

Predicted to react with a broad-range of mammalian species based on 100% sequence homology.
The Tyr1022/1023 designation is according to the JAK1 sequence originally published by Wilks, A.F., et al. (1991). Mol. Cell. Biol. 11(4):2057-2065 (PMID 1848670; NCBI accession: AAA36527). Numbering of the target pTyr residues varies depending on the species and source of the sequence. For example, target pTyr residues this rabbit polyclonal detects correspond to pTyr1034/pTyr1035 and pTyr1033/pTyr1034, respectively, of the human and mouse JAK1 sequence reported by UniProt (P23458 and P52332, respectively), or pTyr1033/pTyr1034 of bovine, canine, and rat JAK1 (NP_001193463 and NP_445918.1), pTyr1021/pTyr1022 of porcine JAK1 (NP_999279).

Immunogen

KLH-conjugated phosphopeptide corresponding to the epitope reqeion sequence containing the two phosphorylated target tyrosine residues.

Application

Detect JAK1 Tyrosine 1022 and Tyrosine 1023 phosphorylation using this rabbit polyclonal Anti-phospho-JAK1 (Tyr1022/1023), Cat. No. 07-849-I, validated for use in Dot Blot and Western Blotting
Dot Blot Analysis: A 1:1,000 dilution of this antibody detected 0.156-20 ng of the immunogen phosphopeptide, but not the corresponding non-phosphorylated peptide.
Research Category
Signaling

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:200 dilution from a representative lot detected Tyr1022/1023-phosphorylated JAK1 in 10 µg of HepG2 cell lysate.

Target description

~130 kDa observed. 133.3 kDa (human JAK1) calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 07-849

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service